Learn More
Tempshield™ Cryo-Industrial Gauntlet Fabric Gloves
Click to view available options
Size:
Medium
Large
X-Large
Dimensione della confezione:
1 paio
Descrizione
- Durable industrial safety gloves for protection of hands and arms when working in hazardous, ultra-cold environments on heavy-duty applications
- Designed to provide a high level of thermal protection, abrasion resistance, flexibility and dexterity
- Manufactured from multi-layered, state-of-the-art materials
- Designed to wick away moisture for comfort when being used for extended periods of time
- Not intended for actual immersion in liquid nitrogen or other cryogenic liquids
Specifica
Specifica
| Colore | Blue |
| Materiale | Fabric |
| Powdered | Powder Free |
| Product Type | Cryogenic Gauntlet |
| Size | Large |
| Sterilità | Non-sterile |
| Lunghezza (sistema metrico) | 430 to 475 mm |