Learn More
Epredia™ Cryotome™ Cryostat Cryocassettes, 25mm dia. Crosshatched
Sfoglia Tutto Epredia Prodotti
Click to view available options
Product Type:
10 mm Circular Cryocassette
Annular 25mm Cryocassette
Annular 35mm Cryocassette
Anti Roll Plate
Crosshatched 25mm Cryocassette
Crosshatched 40mm Cryocassette
Cryobar Cover
Debris Tray
Glass Anti-Roll Plate
Heat Extractor, Double-weight
Heat Extractor, Standard
Knife Guard
Narrow Brush
Narrow Shelf
Tool Shelf
Wide Brush
Wide Shelf
Da utilizzare con (apparecchiatura):
FSE/FE
FSE/FE/SME/E
Dimensione della confezione:
Pezzo
pezzi 4
pezzi 6
Descrizione
- Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
- Freezing time is shortened if freezing stage and tissue are placed on active Peltier
- Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples
Specifica
Specifica
| Descrizione | Knife Guard |
| Product Type | Knife Guard |
| Da utilizzare con (apparecchiatura) | FSE/FE/SME/E |