missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CTCF Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals™ NB500-175PE
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CTCF Polyclonal antibody specifically detects CTCF in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifica
| CTCF | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| PE | |
| 11 zinc finger transcriptional repressor, 11-zinc finger protein, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein), CTCFL paralog, transcriptional repressor CTCF | |
| The immunogen recognized by this antibody maps to a region between residues 150 and 200 of human CCCTC-binding factor using the numbering given in entry NP_006556.1 (GeneID 10664). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 10664 | |
| Store at 4°C in the dark. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto