missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTCF Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA579087
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell, human SiHa whole cell, human HepG2 whole cel, rat brain tissue, mouse brain tissue. ICC/IF: U2OS cell. Flow: MCF-7 cell.
Transcriptional factor CTCF is a ubiquitously expressed 11-zinc finger domain containing, DNA-binding nuclear phosphoprotein that is involved in multiple aspects of normal gene regulation including transcriptional repression and activation, gene silencing, chromatin insulation and regulation of imprinted sites. CTCF uses different combinations of ZF domains and interacts with the CCCTC motif on the DNA. It can bind to HAT- and HDAC-containing complexes thereby influencing transcriptional activation or repression respectively. It is a multifunctional protein involved in the transactivation of APPB gene, silencing of c-myc gene, and insulation of human beta-globin and DMI myotonic dystrophy locus and imprinting control of IGF2 and H19 regions. CTCF has also been shown to regulate the transcription of Pax6 and IRAK2 promoter thereby opening a therapeutic avenue. Mutations in this gene have been associated with invasive breast cancers, prostate cancers and Wilms' tumors.
Specifica
| CTCF | |
| Polyclonal | |
| Unconjugated | |
| CTCF | |
| 11 zinc finger transcriptional repressor; 11-zinc finger protein; AW108038; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); Ctcf; ctcf protein; CTCFL paralog; im:7152666; MRD21; OTTHUMP00000197610; Transcriptional repressor CTCF; Unknown (protein for IMAGE:8125283) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10664, 13018, 83726 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P49711, Q61164, Q9R1D1 | |
| CTCF | |
| E.coli-derived human CTCF recombinant protein (Position: K521-R727). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto