Learn More
CTIF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-83007
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CTIF Polyclonal specifically detects CTIF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifica
| Immunocytochemistry, Immunofluorescence | |
| CBP80/20-dependent translation initiation factor, KIAA0427Gm672 | |
| Human, Mouse |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Affinity Purified |