Learn More
Gibco™ CTS™ Detachable Dynabeads™ CD8
Descrizione
Gibco CTS Detachable Dynabeads CD8 are intended for ex vivo isolation of CD8+ T cell subsets. Combined with CTS Detachable Dynabeads Release Buffer (Cat. No. A5588301, A5588302, or A5588303), it is a first-of-its-kind cGMP cell selection technology with an active release mechanism for process development, clinical trial, and commercial manufacturing use. This product is also commonly used with CTS Detachable Dynabeads CD4 (Cat. No. A56994).
Features of CTS Detachable Dynabeads CD8 include:
- Empower users with process flexibility by allowing active release of beads from target cells at any time
- Optimize target cell purity and yield thanks to the active release mechanism that gives users more control
- Provide an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
- Enable consistent performance in light of biological variability within the starting material
- Suitable for both autologous and allogeneic cell therapy manufacturing processes
These Detachable Dynabeads are meant for direct use with CTS Detachable Dynabeads Release Buffer, which has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of the beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process.
Based on a recommended 4-to-1 bead-to-target-cell ratio (not total nucleated cell number), one 15 mL vial can theoretically isolate up to 1.5 x 109 CD8+ T cells.
CTS Detachable Dynabeads also include a Drug Master File (DMF) to support cross-referencing in regulatory submissions prior to clinical trial and commercial manufacturing use.
Specifica
Specifica
| Concentrazione | 4 x 108 beads/mL |
| Content And Storage | CTS™ Detachable Dynabeads™ CD8, 15 mL. Store at 2°C to 8°C with vial upright to keep beads in suspension. |
| Colore | Brown |
| Materiale | Polystyrene |
| Product Type | Cell Isolation Reagent |
| Da utilizzare con (apparecchiatura) | CTS™ DynaCellect™ Magnetic Separation System |
| Linea di prodotti | CTS Detachable Dynabeads |
| Purity or Quality Grade | GMP-grade |
| Quantità | 15 mL vial |
| Shipping Condition | Ships at 2°C to 8°C with wet (not frozen) gel packs. Vial must remain upright to keep beads in suspension. |
| Vedi altri risultati |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.