Learn More
Sartorius Cubis™ II Analytical Balance, MCE User Interface
Sfoglia Tutto Sartorius Prodotti
Click to view available options
:
Capacity: 220 g; Draft shield U - Manual glass
Descrizione
- Monolithical weighing system
- Integrated climate sensors
- Individual sample holders
- Error-free operation
- Individual QApp workflows
- Motorized auto-leveling
- Full end-to-end data integrity
- 21 CFR Part 11 compliance
- Integrated audit-trail
- State-of-the-art user management
- Excellent service support
- Integrated status center
- Integrated service functions
- Preventive maintenance on accredited standards
- TFT touch screen operation with readable display and easy to operate user interface
- Software Factory installed basic set of essential weighing applications
- Operation Activated by touch key, touch-free using IR sensor
(draft shield M), or gesture sensor (optional) learning capability
Specifica
Specifica
| Certificazione/Conformità | 21 CFR PART 11 17025 USP 41 |
| Tipo di collegamento | D-sub connector, 9-pin |
| Descrizione | Draft shield U: Manual glass |
| Capacità | 220 g |
| Tipo di display | TFT Color Touchscreen |
| Leggibilità | 0.1 mg |
| Include | Analytical balance with manual draft shield |
| Materiale | Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim |
| Modello | Electronic analytical balance |
| Tempo di stabilizzazione | 1 sec. |
| Vedi altri risultati |