Learn More
Sartorius Cubis™ II Precision Lab Balance
Sfoglia Tutto Sartorius Prodotti
Click to view available options
Capacità:
1200 g
14,200 g
2200 g
32,200 g
320 g
3200 g
4200 g
5200 g
620 g
6200 g
8200 g
Modello:
CEU Electronic Toploading Balance
Standard Electronic Toploading Balance
Leggibilità:
1 mg
10 mg
10 mg, 100 mg
10, 20, 50 mg
100 mg
Include:
Draft Shield E, QP99
Draft Shield F, Draft Shield R, QP99
Draft Shield O
Draft Shield O, QP1
Draft Shield O, QP99
Draft Shield R
Draft Shield R, QP99
Draft Shield R,QP99
Dimensione della confezione:
1 unità
Pezzo
Descrizione
The New Generation of Modular Premium Laboratory Balances
- Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry.
- One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series.
- All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems.
- These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement.
- It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Specifica
Specifica
| Certificazione/Conformità | Balance with Type Approval Certificate for China |
| Tipo di collegamento | D-sub connector, 9-pin |
| Capacità | 320 g |
| Tipo di display | Color Touchscreen |
| Leggibilità | 1 mg |
| Include | Draft Shield E, QP99 |
| Materiale | Aluminum, plastic PBT, Optiwhite float glass and stainless steel |
| Modello | CEU Electronic Toploading Balance |
| Tempo di stabilizzazione | 1 sec. |
| Requisiti elettrici | 100 – 240 VAC (±10%), 50 - 60 Hz, 1.0 A |