Learn More
Thermo Scientific™ Culti-Loops™ Actinomyces viscosus ATCCC™ 15987™
Codice prodotto. 12918828
Click to view available options
Quantità:
5 Loops/Pk.
Dimensione della confezione:
pezzi 5
Codice prodotto. 12918828
Marca: Thermo Scientific™ R4600510
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifica
ATCC® 15987TM*
Certificazione/Conformità Culti-Loops are CE Marked.
Descrizione Actinomyces viscosus
Product Type Single Organism
Target Organism Class Gram-positive Rods
Test Type Qualitative QC
Tipo di imballaggio 5 Loops/Pack
Quantità 5 Loops/Pk.
Strain Designations T-6
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.