Learn More
Culti-Loops™ API™ Staph QC Set
Descrizione
Thermo Scientific™ Culti-Loops™ API Staph Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Staphylococcus capitis subsp. capitis ATCC™ 35661™†
Staphylococcus lentus ATCC™ 700403™†
Staphylococcus xylosus ATCC™ 700404™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
Specifica
Specifica
| ATCC® | 35661TM* |
| Descrizione | API Staph QC Set/3 |
| Formato | Culti-Loops |
| Product Type | Culti-Loops QC Set |
| Target Organism Class | Organism Sets |
| Test Type | Qualitative QC |
| Quantità | 4 Loops/Pack |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.