Learn More
Thermo Scientific™ Culti-Loops™ Clostridium baratii ATCC™ 27638™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Marca:  Thermo Scientific™ R4601615
Codice prodotto. 12938918
  • 288.00€ / pezzi 5
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
27638TM*
Culti-Loops
Anaerobes
5 Loops/Pack
VPI 4443
Clostridium barati
Single Organism
Qualitative QC
5 Loops/Pk.
Offerte speciali
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.