Learn More
Thermo Scientific™ Culti-Loops™ Kirby Bauer Susceptibility QC Set
Codice prodotto. 15279267
Click to view available options
Quantità:
5 Loops/Pack
Dimensione della confezione:
Pezzo
Codice prodotto. 15279267
Marca: Thermo Scientific™ R4651101
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ Kirby Bauer Susceptibility Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
(Mueller Hinton) 10 Loops/Organism.
Escherichia coli ATCC™ 25922™†
Pseudomonas aeruginosa ATCC™ 27853™†
Staphylococcus aureus subsp. aureus ATCC™ 25923™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifica
ATCC® 25922TM*
Descrizione CLIA '88 Compliance Kirby Bauer Susceptibility QC Set/3
Formato Culti-Loops
Product Type Culti-Loops QC Set
Target Organism Class Organism Sets
Test Type Qualitative QC
Quantità 5 Loops/Pack
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.