Learn More
Thermo Scientific™ Culti-Loops™ Neisseria perflava ATCC™ 14799™
Codice prodotto. 12935773
Click to view available options
Quantità:
5 Loops/Pack
Dimensione della confezione:
pezzi 5
Codice prodotto. 12935773
Marca: Thermo Scientific™ R4607046
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifica
ATCC® 14799TM*
Descrizione Neisseria perflava ATCC™ 14799™
Formato Culti-Loops
Product Type Culti-Loop
Target Organism Class Fastidious
Test Type Qualitative QC
Quantità 5 Loops/Pack
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.