Learn More
Thermo Scientific™ Culti-Loops™ Nocardia asteroides CL 11014
Codice prodotto. 12945893
Click to view available options
Quantità:
5 Loops/Pack
Dimensione della confezione:
pezzi 5
Codice prodotto. 12945893
Marca: Thermo Scientific™ R4609010
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifica
ATCC® 11014TM*
Descrizione Nocardia asteroides CL 11014
Formato Culti-Loops
Product Type Culti-Loop
Target Organism Class Gram-positive Rods
Quantità 5 Loops/Pack
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.