missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CY 208-243, Tocris Bioscience™
Click to view available options
Quantità:
10 mg
50 mg
Descrizione
Centrally active dopamine D1 receptor agonist, selective over D2 receptor sites. Stimulates adenylate cyclase in rat striatal homogenates with an EC50 of 125 nM. Unlike SKF 38393 (Cat. No. 0922), it exerts antiParkinsonian activity in animal models.
Specifica
Materiale o nome chimico | CY 208-243 |
Quantità | 50 mg |
Peso formulazione | 274.36 |
Percent Purity | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'