Learn More
Cyanoacetamide, 99%, Thermo Scientific Chemicals
Descrizione
Cyanoacetamide was used in spectrofluorimetric method for the determination of some antihistaminic H1 receptor antagonist drugs such as ebastine, cetirizine dihydrochloride and fexofenadine hydrochloride. It may be used in fluorometric determination of 3,4-dihydroxyphenylalanine. It was used as post column fluorigenic derivatizing agent for the bio-analysis and pharmacokinetics of chitosan ester in rabbit serum. Cyanoacetamide is the starting reagent for vitamin B6 synthesis. It reacts with reducing carbohydrates in borate buffer to give intense fluorescence andis useful for the automated analysis of carbohydrates as borate complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyanoacetamide was used in spectrofluorimetric method for the determination of some antihistaminic H1 receptor antagonist drugs such as ebastine, cetirizine dihydrochloride and fexofenadine hydrochloride. It may be used in fluorometric determination of 3,4-dihydroxyphenylalanine. It was used as post column fluorigenic derivatizing agent for the bio-analysis and pharmacokinetics of chitosan ester in rabbit serum. Cyanoacetamide is the starting reagent for vitamin B6 synthesis. It reacts with reducing carbohydrates in borate buffer to give intense fluorescence andis useful for the automated analysis of carbohydrates as borate complexes.
Solubility
Soluble in water.
Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
Specifica
Materiale o nome chimico | Cyanoacetamide |
Punto di fusione | 118°C to 122°C |
Punto d'infiammabilità | 215°C (419°F) |
Quantità | 500 g |
Beilstein | 878221 |
Indice di Merck | 14,2688 |
Informazioni di solubilità | Soluble in water. |
Peso formulazione | 84.08 |
Percent Purity | 99% |
RUO – Research Use Only