missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYR61 Polyclonal Antibody, DyLight™ 550

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA522766
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody is predicted to react with rat and mouse based on 88% sequence homology.
CYR61 is a secreted heparin binding protein, encoded by a growth factor inducible immediate-early gene that associates with the extracellular matrix and connective tissue. CYR61 is a member of a distinct family of angiogenic and vasculogenic regulators designated CCN proteins, which includes connective tissue growth factor (CTGF) and the mouse CYR61 homolog, Fisp12. As an angiogenic inducer, CYR61 binds to the cell surface receptor integrin alphavbeta3, where it then stimulates cell adhesion and migration and promotes DNA synthesis of human vascular endothelial cells. Expression of CYR61 is elevated during vessel growth, wound healing and chondrocyte differentiation. CYR61 is also detected in a wide variety of tumors as it induces tumor growth and functions as a marker of tumor progression.
Specifica
| CYR61 | |
| Polyclonal | |
| DyLight 550 | |
| CCN1 | |
| 3CH61; AI325051; CCN family member 1; CCN1; cellular communication network factor 1; Cyr61; cysteine rich angiogenic inducer 61; cysteine rich protein 61; cysteine-rich angiogenic inducer 61; cysteine-rich heparin-binding protein 61; cysteine-rich, angiogenic inducer, 61; cysteine-rich, anigogenic inducer, 61; GIG1; IBP-10; IGF-binding protein 10; IGFBP10; IGFBP-10; insulin-like growth factor binding protein 10; insulin-like growth factor-binding protein 10; intermediate early; protein CYR61; Protein GIG1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100339657, 16007, 3491 | |
| 4°C, store in dark | |
| Liquid |
| Immunohistochemistry (Paraffin), Neutralization, Western Blot, Immunocytochemistry | |
| 0.58 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| O00622, P18406 | |
| CCN1 | |
| A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rabbit | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto