missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 3A4/3A5 Antibody (F24P2B10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-50208
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Cytochrome P450 3A4/3A5 Monoclonal antibody specifically detects Cytochrome P450 3A4/3A5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifica
| Cytochrome P450 3A4/3A5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Albendazole monooxygenase, Albendazole sulfoxidase, CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, Cytochrome P450 3A3, cytochrome P450 3A4, Cytochrome P450 HLp, Cytochrome P450 NF-25, cytochrome P450, family 3, subfamily A, polypeptide 4, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4, Cytochrome P450-PCN1, EC 1.14.13.32, EC 1.14.13.67, EC 1.14.13.97, EC 1.14.14.1, glucocorticoid-inducible P450, HLP, MGC126680, NF-25, Nifedipine oxidase, P450C3, P450-III, steroid inducible, P450PCN1, Quinine 3-monooxygenase, Taurochenodeoxycholate 6-alpha-hydroxylase | |
| Ovalbumin-conjugated synthetic peptide ESRDGTVSGA | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1576 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| F24P2B10 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto