Learn More
Epredia™ DAB Quanto Detection System
Sfoglia Tutto Epredia Prodotti
Click to view available options
Quantità:
1 L
125 ml di substrato DAB Quanto, 2 ml di cromogeno DAB Quanto
60 mL DAB Quanto Substrate, 2 mL DAB Quanto Chromagen
Dimensione della confezione:
125mL
1L
60mL
Descrizione
DAB Quanto has been validated in combination with both UltraVision LP and UltraVision Quanto and provides four major benefits.
- Strong staining—ideal to quantifying cells and protein expression
- Less mixing—reducing personnel time and stress
- Less disposal—cuts down waste
- Lower risk of mixing errors—supports higher staining consistency
- Supplied in a two-component form consisting of a DAB Quanto Chromogen and DAB Quanto Substrate
- Standard working dilution is 30μL of DAB Quanto Chromogen per 1mL of DAB Quanto Substrate
- Use of liquid components reduces some risks associated with the handling of powders or tablets
Specifica
Specifica
| Tipo di cromogeno | DAB Quanto Chromagen |
| Quantità | 1 L |
| Tipo di substrato | Dab Quanto Subrate |