missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DAPT

Click to view available options
Quantità:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
Descrizione
DAPT is a γ-secretase inhibitor. DAPT reduces Aβ40 and Aβ42 levels in human primary neuronal cultures (IC50 values are 115 and 200 nM for total Aβ and Aβ42 respectively) and in brain extract, cerebrospinal fluid and plasma in vivo. DAPT has no effect on APPα and APPβ levels. DAPT blocks Notch signaling in hybrid human-mouse fetal thymus organ culture (FTOC) and causes ESCs to commit to neuronal differentiation. DAPT can be used in a small molecule cocktail to derive cortical neurons from hPSCs and to maintain hepatocytes in culture. DAPT also promotes the formation of cone photoreceptors in retinal organoids.
For more information about how DAPT may be used, see our protocols: Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts, Generating Midbrain Dopaminergic Neurons from hPSCs .
DAPT synthesized to Ancillary Material Grade also available.
Specifica
Specifica
| Materiale o nome chimico | DAPT |
| Quantità | 10 mg |
| Peso formulazione | 432.46 |
| Purezza | >99% |
| Peso molecolare | 432.468 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto