Learn More
Thermo Scientific™ Data Output Accessories for Environmental Chambers and Incubators
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Modello:
Chart Paper
Dual Pen
Single Pen
Tipo:
7 Day Chart Recorder
Chart Paper
Installazione:
Customer Installed
Factory Installed
Dimensione della confezione:
Pezzo
Descrizione
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifica
Specifica
| Descrizione | Chart paper for single pen chart recorders |
| Da utilizzare con (apparecchiatura) | Environmental Chambers and Incubators |
| Lighting | LED |
| Temperature Range (Metric) | -10°C to +70°C |
| Tipo | Chart Paper |
| Installazione | Customer Installed |
| Modello | Chart Paper |