Learn More
Thermo Scientific™ Data Output Accessories for Environmental Chambers and Incubators
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Modello:
Chart Paper
Dual Pen
Single Pen
Tipo:
7 Day Chart Recorder
Chart Paper
Installazione:
Customer Installed
Factory Installed
Dimensione della confezione:
Pezzo
Descrizione
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifica
Specifica
| Descrizione | Dual Pen, 6 in. 7 Day Chart Recorders; 0° to 60°C (32° to 140°F) |
| Da utilizzare con (apparecchiatura) | Environmental Chamber (3948 and 3907) |
| Lighting | LED |
| Temperature Range (Metric) | 0°C to 60°C |
| Tipo | 7 Day Chart Recorder |
| Installazione | Customer Installed |
| Modello | Dual Pen |