Learn More
Lenz Laborglasintrumente Delivery Adapters, Bent, with Ventilation
Click to view available options
N. giunto rastremato standard:
Socket and Cone NS 14/23
Socket and Cone NS 19/26
Socket and Cone NS 24/29
Socket and Cone NS 29/32
Dimensione della confezione:
Pezzo
Descrizione
- Delivery adapters
- Bent (105°)
- For highly boiling liquids
- With ventilation
- With NS socket and NS cone
- With drain tip
- Made of DURAN™ tubing
Specifica
Specifica
| Materiale | Duran Glass |
| N. giunto rastremato standard | Socket and Cone NS 29/32 |