Learn More
Chemsplash Delta Coverall (Sterile Irradiated)

Descrizione
Chemsplash Delta 67 Coverall
- 67GSM Microporous Laminate Fabric
- Sterile Irradiated, Double Bagged and Detex Sticker
- Low Linting Knitted Cuffs with Thumb Loops
- Elasticated Three Piece Hood
- Two Way Zip
- Adhesive Zip Flap
- Elasticated Back and Ankles
- Reinforced Bound Seam at Crotch Area
- Non Linting and Anti-Static
- Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Delta 67 Coverall is made from soft, comfortable 67gsm microporous laminate fabric, meeting Cat III Type 5B/6B. It is Sterile Irradiated, double-bagged, and comes with a Detex sticker for identification. The coverall is compliant with EN1149-5:2008 (Antistatic) and EN1073-2 (Nuclear Particle). Chemsplash Delta is ideal for prolonged wear, offering improved breathability while maintaining an effective hazard barrier. This coverall is suitable for a wide range of applications, with an elasticated back, hood, cuffs, ankles, and reinforced seams for added durability. It conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9.
Specifica
Specifica
| Product Type | Cleanroom Coveralls |
| Size | Medium |
| Sesso | Unisex |
| Materiale | Microporous Fabric |
| Colore | White |
| Monouso | Single-use |
| Stile vita | Elastic |
| Ankle Style | Elastic |
| Stile polso | Knit, Thumb Loop |
| Stile collo | Con cappuccio |
| Vedi altri risultati |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.