Learn More
Chemsplash Delta Coverall (Sterile Irradiated)
GREENER_CHOICE
Codice prodotto. 30058401
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 25
Codice prodotto. 30058401
Marca: Chemsplash 2766.400.007
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Delta 67 Coverall - Sterile Irradiated - is made from our soft comfortable 67 gsm microporous laminate fabric, meeting Cat III Type 5B/6B.
Chemsplash Delta 67 Coverall
  • 67GSM Microporous Laminate Fabric
  • Sterile Irradiated, Double Bagged and Detex Sticker
  • Low Linting Knitted Cuffs with Thumb Loops
  • Elasticated Three Piece Hood
  • Two Way Zip
  • Adhesive Zip Flap
  • Elasticated Back and Ankles
  • Reinforced Bound Seam at Crotch Area
  • Non Linting and Anti-Static
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Delta 67 Coverall is made from soft, comfortable 67gsm microporous laminate fabric, meeting Cat III Type 5B/6B. It is Sterile Irradiated, double-bagged, and comes with a Detex sticker for identification. The coverall is compliant with EN1149-5:2008 (Antistatic) and EN1073-2 (Nuclear Particle). Chemsplash Delta is ideal for prolonged wear, offering improved breathability while maintaining an effective hazard barrier. This coverall is suitable for a wide range of applications, with an elasticated back, hood, cuffs, ankles, and reinforced seams for added durability. It conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9.
Specifica
Product Type Cleanroom Coveralls
Size X-Large
Sesso Unisex
Materiale Microporous Fabric
Colore White
Monouso Single-use
Stile vita Elastic
Ankle Style Elastic
Stile polso Knit, Thumb Loop
Stile collo Con cappuccio
Hood Type 3-panel
Tipo di chiusura Ties
Tipo di giuntura Stitched
Sterilità Sterile
Resistenza Liquid
Senza lattice
N. di tasche None
Cleanroom Class Class 100-100,000 (ISO 5-8)