Learn More
Dengue Virus 2 Envelope Antibody (144), FITC , Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-05828F
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Dengue Virus 2 Envelope Monoclonal antibody specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for ELISA
Specifica
| Dengue Virus 2 Envelope | |
| Monoclonal | |
| FITC | |
| Envelope protein E | |
| Produced in mouse immunized with recombinant Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E/ Envelope Protein (Domain III) (AAC59274.1; Met578-Gln680). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 144 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |