Learn More
Alla France™ Idrometro di densità
Click to view available options
Intervallo di gravità specifica:
0.7 to 0.750 g/mL
0.750 to 0.800 g/mL
0.800 to 0.850 g/mL
0.850 to 0.900 g/mL
0.900 to 0.950 g/mL
0.950 to 1.000 g/mL
1.000 - 1.050 g/ml
1.050 - 1.100 g/ml
1.100 to 1.150 g/mL
1.150 to 1.200 g/mL
1.200 to 1.250 g/mL
1.250 to 1.300 g/mL
1.300 to 1.350 g/mL
1.350 to 1.400 g/mL
1.400 to 1.450 g/mL
Dimensione della confezione:
Pezzo
Descrizione
- Density Hydrometers (g/mL)
- ISO, BS, DIN, NF... MRA Traceable
- General Purpose at 20°C
- Reading below meniscus
- Systematically delivered with a Traceability and Conformity Certificate
- On request, can be supplied with "EU" Traceable Calibration Certificate or "EO" Official Calibration Certificate
- Calibrated according to the following surface tensions:
- from 0.600 to 1.000 g/mL= 16 at 35 mN/m (low tension)
- from 1.000 to 1.300 g/mL= 55 mN/m (medium tension)
- from 1.300 to 2.000 g/mL= 75 mN/m (high tension)
Specifica
Specifica
| Quantità | 1Each |
| Tipo | Density Hydrometer |
| Accuracy | ±0.0005g/mL |
| Lunghezza (sistema metrico) | 335 mm |
| Intervallo di gravità specifica | 0.750 to 0.800 g/mL |