Learn More
Density Standards (15 °C), Paragon Scientific™
Click to view available options
Densità:
0.6654 g/mL (15°C)
0.7183 g/mL (15°C)
0.7807 g/mL (15°C)
0.8111 g/mL (15°C)
0.8494 g/mL (15°C)
0.8648 g/mL (15°C)
0.8811 g/mL (15°C)
0.9413 g/mL (15°C)
0.9823 g/mL (15°C)
1.0248 g/mL (15°C)
1.0687 g/mL (15°C)
1.1280 g/mL (15°C)
1.1962 g/mL (15°C)
1.2829 g/mL (15°C)
1.6300 g/mL (15°C)
Dimensione della confezione:
60mL
Descrizione
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols.
- Supplied in tamper evident packaging (please refer to standard for shelf life).
- Manufactured in the United Kingdom and available from stock for immediate dispatch
Specifica
Specifica
| Densità | 0.8811 g/mL (15°C) |
| Quantità | 60 mL |
Nominal values quoted