Learn More
Invitrogen™ Dermatophagoides pteronyssinus Recombinant Rabbit Monoclonal Antibody (068)
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA560556
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Dermatophagoides pteronyssinus Recombinant Monoclonal Antibody for ELISA
Specifica
| Dermatophagoides pteronyssinus | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Recombinant Dermatophagoides pteronyssinus Der p 1 protein. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| 068 | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| RUO | |
| Insect | |
| Antibody | |
| IgG |