missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA561923
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: PDGSLSQAAM MQSALAKERR ELKQAQREAE MDSIPMGLNK HWVDPLPDAE GRQIAANMRG IGMMPNDIPE WKKHAFGGNK ASYGKKTQMS ILEQRESLP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene is a member of the DEAH box polypeptide family. The encoded protein contains the DEAH (Asp-Glu-Ala-His) motif which is characteristic of all DEAH box proteins, and is thought to function as an ATP-dependent RNA helicase that regulates the release of spliced mRNAs from spliceosomes prior to their export from the nucleus. This protein may be required for the replication of human immunodeficiency virus type 1 (HIV-1). Alternative splicing results in multiple transcript variants. [provided by RefSeq, Oct 2014].
Specifica
| DHX8 | |
| Polyclonal | |
| Unconjugated | |
| Dhx8 | |
| ATP-dependent RNA helicase DHX8; DDX8; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 8; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 8 (RNA helicase); DEAH (Asp-Glu-Ala-His) box polypeptide 8; DEAH box protein 8; DEAH-box helicase 8; DHX8; HRH1; LOW QUALITY PROTEIN: ATP-dependent RNA helicase DHX8; LRRGT00035; mDEAH6; mKIAA4096; PRP22; PRPF22; RNA helicase; RNA helicase HRH1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1659 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14562 | |
| Dhx8 | |
| Recombinant protein corresponding to Human DHX8. Recombinant protein control fragment (Product #RP-101103). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto