Learn More
Diclomezine 100 μg/mL in Acetone, Dr. Ehrenstorfer
Codice prodotto. 30311497
Click to view available options
Quantità:
1 mL
Codice prodotto. 30311497
Marca: Dr. Ehrenstorfer DREXA12545000AC
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Diclomezine
CAS 62865-36-5
Nomi componenti o analita Diclomezine
Shipping Condition Room Temperature
Formula molecolare C11 H8 Cl2 N2 O
Concentrazione 100 μg/mL
Certificazione/Conformità ISO/IEC 17025
Sinonimo 6-(3,5-Dichloro-4-methylphenyl)-3(2H)-pyridazinone, Diclomezine, Monguard
InChI Formula InChI=1S/C11H8Cl2N2O/c1-6-8(12)4-7(5-9(6)13)10-2-3-11(16)15-14-10/h2-5H,1H3,(H,15,16)
Tipo di soluzione Single Solution
SMILES Cc1c(Cl)cc(cc1Cl)C2=NNC(=O)C=C2
IUPAC Name 3-(3,5-dichloro-4-methylphenyl)-1H-pyridazin-6-one
Solvent or Matrix Acetone
Industry Type Environmental Testing
Molecular Weight (g/mol) 255.1
Quantità 1 mL
Peso formulazione 254.0014 g/mol