Learn More
Cole-Parmer™ Digi-Sense™ Traceable™ Triple-display Clock/Timer with Calibration
Sfoglia Tutto Cole-Parmer ProdottiBeschreibung
- Get accurate, reliable measurements with included NIST-traceable calibration.
- Timer displays time of day, or time remaining in any of the three timer chanels.
- Each channel can be used either simultaneously or independently.
- Memory recalls previous time set.
- Display allows simultaneous up and down timing on different channels with unlimited time-outs.
- Alarm tones are unique for each channel and have adjustable volume.
- Flashing LCD indicates alarm condition.
- Traceable™ products are calibrated to NIST-traceable standards and ready to use upon receipt.
- Each product goes through an intense vetting process before it can carry the Traceable name and it includes NIST-traceable certification.
- This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately.
- NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
Spezifikation
Spezifikation
| Intervallo di tempo | 99 hr. 59 min. 59 sec. |
| Alarms | 1 min. Continuous Beep and Visible |
| Accuracy | ±0.001% |
| Risoluzione | 1 sec. |
| Display | LCD |
| Descrizione | Traceable™ triple-display clock/timer with calibration |
| N. di canali | 3 |
| Larghezza (sistema metrico) | 8.3 cm |
| Altezza (sistema imperiale) display | 3/8 in. |
| Altezza (sistema metrico) | 7.6 cm |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?