Learn More
Gilson™ Termoblocco Digitale a 2 posizioni, spina EU
Sfoglia Tutto Gilson Prodotti
Click to view available options
Tipo di spina:
EU
Dimensione della confezione:
Pezzo
Descrizione
Accurate temperature delivery
- The Digital Drybath Series offers convenient digital temperature and time controls with built-in calibration functionality, improving the reliability and reproducibility of your reactions without external thermometers or timers.
- The Digital Drybath Series provides a temperature range of +5°C−150°C that is adjustable in 0.1°C increments.
- The timer can be set for up to 999 minutes.
Uniform temperature across all samples
- The Digital Drybath Series accommodates a wide variety of high-grade aluminum blocks that are precisely engineered to the shape of 0.5, 1.5, 2.0, 15, and 50 mL tubes, providing uniform temperature delivery to all your samples and improving reliability across reactions.
- Blocks are sold separately; custom blocks are available upon request.
- Each dry bath is supplied with an insulated block lifter to remove hot blocks.
Specifica
Specifica
| Tipo di spina | EU |
| Temperature Range (Metric) | Ambient +5° to 150°C |
| N. di blocchi | 2 |
| Precisione termica | ±0.2°C |
| Dimensioni (L x P x A) | 22 x 26 x 8 cm |
| Lunghezza (sistema metrico) | 22 cm |
| Larghezza (sistema metrico) | 26 cm |
| Altezza (sistema metrico) | 8 cm |
| Peso (sistema imperiale) | 5.5 lb. |
| Peso (sistema metrico) | 2.5 kg |
| Vedi altri risultati |