missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIP2C Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA566248
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: KKRSKLIGAYL PQPPRVDQAL PQERRAPVTP SSASRYHRRR SSGSRDERY Highest antigen sequence identity to the following orthologs - mouse 100%, rat 68%.
This gene encodes a member of the disco-interacting protein homolog 2 family. The protein shares strong similarity with a Drosophila protein which interacts with the transcription factor disco and is expressed in the nervous system.
Specifica
| DIP2C | |
| Polyclonal | |
| Unconjugated | |
| DIP2C | |
| 2900024P20Rik; 9630044M06; DIP2 disco-interacting protein 2 homolog C; DIP2 disco-interacting protein 2 homolog C (Drosophila); DIP2 homolog C; DIP2C; disco interacting protein 2 homolog C; disco-interacting protein 2 homolog C; KIAA0934; mKIAA0934; RGD1560155 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22982 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2E4 | |
| DIP2C | |
| Recombinant Human DIP2C. Recombinant protein control fragment (Product #RP-105126). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto