Learn More
DWK Life Sciences Disposable Culture Tube, from soda-lime glass, straight rim
Codice prodotto. p-7189499 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Capacità (sistema metrico):
4 mL
6 mL
7 mL
8 mL
9 mL
14 mL
15 mL
18 mL
19 mL
22 mL
Diametro (sistema metrico):
10 mm
11.75 mm
12.25 mm
15.5 mm
15.75 mm
Dimensione della confezione:
pezzi 250
pezzi 310
pezzi 500
pezzi 550
Codice prodotto. 15601604
Marca: DWK Life Sciences 231720862
Solo null disponibili
Aggiungi al carrello
Edge
Aggiungi al carrello
Culture tubes are used for growing cultures from liquid media.
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today''s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. In addition to culture tubes made of DURAN™, our product range also includes culture tubes made of soda-lime glass. This is a glass of the third water resistance Class And belongs to the category of soda-lime glasses with a high proportion of alkali and alkaline earth oxides.
  • Culture tubes with straight edges can be closed with metal caps or cotton plugs to allow air exchange
  • The straight rim permits the use of Kapsenberg and metal caps
TRUSTED_SUSTAINABILITY
Specifica
Descrizione 0.55 mm thickness
Capacità (sistema metrico) 6 mL
Lunghezza (sistema metrico) 75 mm
Materiale Vetro sodico calcico
Da utilizzare con Growth of microorganisms; For microbiological work
Quantità 550/Pk.
Autoclavabile Not Autoclavable
Diametro (sistema metrico) 11.75 mm