Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz-Thiele, Bent
Click to view available options
Giunto rastremato standard:
Socket and Cone NS 14/23
Socket and Cone NS 29/32
Capacità (sistema metrico):
25 mL
50 mL
100 mL
Materiale:
Duran Glass
Duran PTFE
Dimensione della confezione:
Pezzo
Descrizione
- Receivers with bent adapter, graduated
- With NS socket and NS cone
- vacuum connection: thread GL18
- NS stopcock with solid glass or PTFE plug
- Made of DURAN™ tubing
Specifica
Specifica
| Include | Bent adapter, graduated, with NS socket and NS cone, vacuum connection: thread GL18 |
| Materiale | Duran Glass |
| Capacità (sistema metrico) | 25 mL |
| Giunto rastremato standard | Socket and Cone NS 29/32 |