missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLL4 Antibody (MHD4-46), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-62229PEATT594
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
DLL4 Monoclonal specifically detects DLL4 in Human samples. It is validated for Flow Cytometry.
Specifica
| DLL4 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| DLL4 | |
| recombinant soluble human DLL4 | |
| 0.1 mL | |
| Primary | |
| The antibody GF-01 reacts with GFAP, the principal marker of astroglial cells in the central nervous system, which is specifically expressed in satellite cells in peripheral ganglia and in non myelinating Schwann cells in peripheral nerves. The GFAP protein runs on gels at ∽55 kDa protein, usually associated with lower Mw bands which are thought to be proteolytic fragments and alternate transcripts from the single gene. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| MHD4-46 | |
| Flow Cytometry | |
| delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 | |
| Mouse | |
| Protein A purified | |
| Neuroscience | |
| 54567.0 | |
| Human | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto