Learn More
DLL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP1-86413
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
DLL4 Polyclonal specifically detects DLL4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CHDLENGLMCTCPAGFSGRRCEVRTSIDACASSPCFNRATCYTDLSTDTFVCNCPYGFVGSRCEFPVGL | |
| 0.1 mL |
Titolo del prodotto
For Research Use Only