missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11713-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.Specifica
| DNAJA1 | |
| Polyclonal | |
| Unconjugated | |
| DNAJA1 | |
| DJ 2, DjA1, DnaJ protein homolog 2, DNAJ2, DNAJA1, hDJ 2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, HSDJ, HSJ 2, HSJ2, HSPF4, Human DnaJ protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15502, 3301 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P31689, P63037 | |
| Dnaja1 | |
| DNAJA1 Fusion Protein Ag2284 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto