Learn More
DNAJA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12236-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The protein encoded by this gene belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain. The product of this gene works as a cochaperone of Hsp70s in protein folding and mitochondrial protein import in vitro.Specifica
| DNAJA2 | |
| Polyclonal | |
| Unconjugated | |
| DNAJA2 | |
| CPR3, DJ3, DJA2, DNAJ, DNAJA2, DNJ3, HIRA interacting protein 4, HIRIP4, PRO3015, RDJ2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10294, 56445, 84026 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35824, O60884, Q9QYJ0 | |
| Dnaja2 | |
| DNAJA2 Fusion Protein Ag2877 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |