missing translation for 'onlineSavingsMsg'
Learn More

DNAJB9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifica

Antigene DNAJB9
Concentrazione 0.25 mg/mL
Applicazioni Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Coniugato Unconjugated
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
16801234
Vedere i documenti
Proteintech
13157-1-AP-20UL
20 μL
145.00€
20µL
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16891224
Vedere i documenti
Proteintech
13157-1-AP-150UL
150 μL
410.00€
150µL
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.
Specifica

Specifica

DNAJB9
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Mouse, Human
P97554, Q9QYI6, Q9UBS3
24908, 27362, 4189
DNAJB9 Fusion Protein Ag3789
Primary
-20°C
DNAJB9
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
DNAJB9, ERdj4, Mdg 1, MDG1, MST049, MSTP049
DNAJB9
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.