missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538712
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The downstream of kinase family (Dok-1-7) are members of a class of "docking" proteins that include the tyrosine kinase substrates IRS-1 and Cas, which contain multiple tyrosine residues and putative SH2 binding sites. Dok-4, Dok-5 and Dok-6 are more similar to each other than to the other Dok family members, and may constitute a subfamily of the DOK genes. Dok-6 is highly expressed in the developing central nervous system. It associates with Ret to transduce Ret-mediated processes such as axonal projection.
Specifica
| DOK6 | |
| Polyclonal | |
| Unconjugated | |
| DOK6 | |
| Docking protein 6, Downstream of tyrosine kinase 6, DOK5L, docking protein 5-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 220164, 623279 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6PKX4, Q2MHE5 | |
| DOK6 | |
| A synthetic peptide derived from the internal region of human DOK6 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto