missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Donkey anti-Goat IgG Fab Secondary Antibody, Biotin
Donkey Polyclonal Secondary Antibody
Marca: Invitrogen™ PA129616
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Goat IgG Fab | |
| Polyclonal | |
| Biotin | |
| Donkey | |
| IgG fraction | |
| RUO | |
| Goat | |
| Secondary Antibody | |
| IgG F(ab) |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA and 0.01% sodium azide; pH 7.2 | |
| Goat IgG whole molecule. | |
| 500 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto