Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody has been used in 2 publications
Marca: Novus Biologicals NB7379
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifica
| IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Goat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 0.5 mg/mL | |
| Western Blot 1:5000-1:50000, ELISA 1:10000-1:100000, Immunohistochemistry 1:200-1:5000, Immunocytochemistry/Immunofluorescence 1:200-1:5000, Immunohistochemistry-Frozen 1:200-1:5000 | |
| Goat IgG-heavy and light chain | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with goat IgG and with light chains common to other goat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. Antiserum was cross adsorbed using chicken, human, mouse, pig, rabbit and rat immunosorbents to remove cross reactive Antibodies. Less than 0.1% cross reactivity to chicken, horse, human, mouse, pig, rabbit and rat IgG was detected. This antibody may cross react with IgG from other species. | |
| Store at 4C. Do not freeze. |
Titolo del prodotto
For Research Use Only