Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
Donkey Polyclonal Antibody
Marca: Novus Biologicals NBP2-60656JF549
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Dot Blot, Fluorophore-linked immunosorbent assay
Specifica
| IgG (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Donkey anti-Goat IgG (H+L) Secondary Antibody was produced by repeated immunization with goat IgG whole molecule in donkey | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Goat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Dot Blot | |
| Janelia Fluor 549 | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |