missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey Anti-Sheep IgG (H+L) PerCP-conjugated Antibody, R&D Systems™

Donkey Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems F0128
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Flow Cytometry.
Specifica
| IgG | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| IGHG1 | |
| Sheep IgG | |
| 100 Tests | |
| Secondary | |
| This donkey anti-sheep IgG (H+L) antibody was purified by sheep IgG (H+L) affinity chromatography and absorbed against human IgG, and human, mouse, bovine and rabbit serum to eliminate cross-reactivity. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| PerCP | |
| Immunoglobulin G, ImmunoglobulinG | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| Sheep | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto