missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey F(ab')2 anti-Rabbit IgG (H+L) Secondary Antibody, PE (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Marca: Novus Biologicals NB7581
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifica
| IgG (H+L) | |
| Polyclonal | |
| PE | |
| Donkey | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Reconstitute to desired concentration. | |
| Store at 4°C in the dark. Do not freeze. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot 1:1000 - 10000, Flow Cytometry 1:100 - 1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:200 | |
| Donkey F(ab')2 anti-Rabbit IgG (H+L) Secondary Antibody [PE] (Pre-adsorbed) was produced by repeated immunization with Rabbit IgG whole molecule in donkey. | |
| 0.25 mg | |
| Adaptive Immunity, Immunology | |
| No reaction was observed against anti-Pepsin, anti-Donkey IgG F(c) or to Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat & Sheep Serum Proteins. | |
| Rabbit | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto