Learn More
Socorex™ Dosys™ Basic Pistol Grip Automatic Syringe
Codice prodotto. 10437092
Click to view available options
Volume (sistema metrico):
0.1 to 0.5 mL
0.3 to 2 mL
0.5 to 5 mL
Precisione:
<0.5% to <0.2%
<0.6% to <0.4%
<0.7% to <0.4%
Dimensione della confezione:
Pezzo
Codice prodotto. 10437092
Marca: Socorex™ 172.0501
Dose with excellent reproducibility using a pistol grip handle automatic syringe. Socorex™ Dosys™ Basic Pistol Grip Automatic Syringe fits either hand comfortably and requires limited effort for efficient work even during long dispensing sessions.
  • Pistol grip handle.
  • Automatic syringe.
  • Without valve system.
  • Luer lock nozzle.
  • Fits either hand.
  • Requires limited effort.
  • High reproducibility due to safety locking nut on a micrometric screw.
  • High-precision stainless steel plunger.
  • Ground glass barrel.
  • Smooth plunger travel and tightness.
  • Glass barrel protected by PVC sleeve (not autoclavable).
  • Easy disassembly for efficient cleaning and parts replacement if necessary.
Specifica
Autoclavabile Autoclavable
Materiale Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Include Syringe, PVC protection sleeve, operating instructions.
Product Type Automatic Syringe, Pistol Grip
Graduazioni 0.05 mL
Modello Dosys™ Basic 172
Quantità 1/ea.
Precisione <0.7% to <0.4%
Volume (sistema metrico) 0.1 to 0.5 mL
Graduated Yes