Learn More
Dräger™ Pompa a soffietto accuro™: Solo pompa
Codice prodotto. 10588931
Cambia vista
Click to view available options
Tipo:
Pompa a soffietto Accuro™
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Dimensione della confezione
10588931 Pompa a soffietto Accuro™ Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10588931 Fornitore Dräger™ N. del fornitore 6400000
  • ONLINE EXCLUSIVE PRICE
    1405.80€
    1213.18€ / pezzi 1
    Risparmia 192.62€
    13% Sconto
Spedizione stimata: 10-12-2026
Requires
Dräger Short-Term Detector Tubes (sold separately)
Ergonomic design allows easy measurement of more than 350 gases—including formaldehyde
Provides a means of direct, onsite measuring of more than 350 gases, vapors, and aerosols. Internal scissors mechanism provides stability so that a uniform, even pump stroke of 100cm3 can be completed.
  • Convenient, built-in stroke counter records strokes performed
  • Recessed counter reset button prevents accidental resetting; a white dot in clear optical window of pump rises to indicate completion of each stroke cycle, preventing incomplete measurements
  • Ergonomic design fits comfortably in one hand
  • Soft rubber grips on pump body make it easier to hold and squeeze
  • Built-in ceramic tube opener adds safety and convenience Corrosion-resistant construction provides years of reliable service
  • Components are easily replaceable and snap together for quick reassembly
  • Use with accessory extension hoses for remote sampling
  • Designed and manufactured using ISO 90001:2008-certified processes
Order Info
Pump dimensions: 6.75L x 1.75W x 3.75 in. H (17.1 x 4.5 x 8.5cm). Net weight: 0.5 lb. (225g). Stroke volume: 100cm3 ±5%. Hardside Kit, Softside Kit and hard case available separately (Cat. Nos. 19-046-750, 17-988-408 and 19-046-749).
Specifica
Tipo Pompa a soffietto Accuro™