Learn More
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug
Codice prodotto. p-7116294
Click to view available options
Capacità (sistema metrico):
100 mL
1000 mL
2000 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 11886184
Marca: Lenz Laborglasinstrumente™ 04473702
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Made of borosilicate glass. Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped, with PTFE plug are used for transferring fluids. The funnel is fitted with a stopcock which allows the flow to be controlled.
  • Conical shape with NS sleeve and NS core
  • With PTFE plug
  • With threaded valve protection
  • Borosilicate glass
Specifica
Capacità (sistema metrico) 100 mL
Materiale Borosilicate Glass
Dimensione del poro 2.5 mm bore
Stopper Material PTFE